Ac-Arg-Leu-Arg-AMC
Supplier: ADIPOGEN CORP MS
AG-CP3-0013-M001
AG-CP3-0013-M005
102988-470EA
76.15
USD
102988-470
102988-472
Ac-Arg-Leu-Arg-AMC
Proteins and Peptides
Fluorogenic tri-peptide substrate for measuring the trypsin-like peptidase activity of the 20S proteasome. It is cleaved exclusively at the amide R-AMC bond to release fluorescent product. Shows low Km and high specific activity. Excitation: 380nm. Emission: 460nm. This substrate is useful for inhibitor screening and kinetic analysis.
- Soluble in 5% acetic acid.