BQ-123 sodium salt ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

Synonyms: Endothelin Receptor A Antagonist BQ-123

AG-CP3-0001-M001 AG-CP3-0001-M005
102988-434EA 145.38 USD
102988-434 102988-436
BQ-123 sodium salt ≥98% (by HPLC)
BQ-123 sodium salt

Potent and selective endothelin receptor A (ETA) antagonist. Suppresses the ET-1-induced decrease of LPL activity. Shows protective effects in ischemic acute renal failure. Neuroprotective. Anti-hypertensive. Inhibits ET-1 receptor binding and blocks Ca2+ mobilization, cellular contraction and MAP kinase activation. Cardiovascular agent. Reduces myocardial infarct size and oxidant injury. Selected Reviews.

Formula: C₃₁H₄₂N₆O₇·Na
MW: 632.7 g/mol
Storage Temperature: Freezer
CAS Number: 136655-57-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White solid.
Solubility Soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR