Asialo-Ganglioside GM1 ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS

Synonyms: Ceramide tetrahexoside

AG-CN2-9008-C500 AG-CN2-9008-M001
102988-428EA 249.23 USD
102988-428 102988-430
Asialo-Ganglioside GM1 ≥98% (by TLC)
Asialo ganglioside GM1

Gangliosides are acidic glycosphingolipids that form lipid rafts in the outer leaflet of the cell plasma membrane, especially in neuronal cells in the central nervous system. They participate in cellular proliferation, differentiation, adhesion, signal transduction, cell-to-cell interactions, tumorigenesis and metastasis. The accumulation of gangliosides has been linked to several diseases. Removing the sialic acid from monosialoganglioside GM1 inhibits its ability to bind cholera toxin. Asialo-ganglioside GM1 does not block the binding of cholera toxin to cellular GM1.

Formula: C₆₂H₁₁₄N₂O₂₃
MW: 1255.6 g/mol (calc. on sphingosine C18:1 and stearic acid)
Storage Temperature: Freezer
CAS Number: 71012-19-6

Order Now

Specification Test Results

Source/Host Semisynthetic.
Purity 98% (TLC)
Solubility Soluble in chloroform:methanol (2:1).
Formulation Lyophilized.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR