Ganglioside GT1b trisodium salt ≥98%
Supplier: ADIPOGEN CORP MS
Synonyms:
GT1b . 3Na
AG-CN2-9006-M001
AG-CN2-9006-M005
102988-420EA
193.85
USD
102988-420
102988-422
Ganglioside GT1b trisodium salt ≥98%
Ganglioside GT1b trisodium salt
Gangliosides are acidic glycosphingolipids that form lipid rafts in the outer leaflet of the cell plasma membrane, especially in neuronal cells in the central nervous system. They participate in cellular proliferation, differentiation, adhesion, signal transduction, cell-to-cell interactions, tumorigenesis and metastasis. The accumulation of gangliosides has been linked to several diseases. Ganglioside GT1b binds botulinum and tetanus neurotoxins and it prevents neurotoxicity of glutamate on neuronal cells.
Formula:
C₉₅H₁₆₂N₅O₄₇·₃Na Storage Temperature: Freezer |
CAS Number:
59247-13-1 |
Specification Test Results
Source/Host | Isolated from bovine brain. |
Purity |
|
Solubility | Soluble in water (micellar aggregates) or chloroform:methanol (2:1). |
Formulation | Lyophilized |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...