Ganglioside GD1a disodium salt ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS

Synonyms: GD1a . 2Na

AG-CN2-9003-M001 AG-CN2-9003-M005
102988-410EA 124.62 USD
102988-410 102988-412
Ganglioside GD1a disodium salt ≥98% (by TLC)
Ganglioside GD1a disodium salt

Gangliosides are acidic glycosphingolipids that form lipid rafts in the outer leaflet of the cell plasma membrane, especially in neuronal cells in the central nervous system. They participate in cellular proliferation, differentiation, adhesion, signal transduction, cell-to-cell interactions, tumorigenesis and metastasis. The accumulation of gangliosides has been linked to several diseases. Ganglioside GD1a is the major ganglioside of the nervous system. It is converted to GM1 by bacterial, viral and mammalian sialidases. It is a differentiation marker for cell growth.

Formula: C₈₄H₁₄₆N₄O₃₉·2Na
MW: 1882.1 g/mol
Storage Temperature: Freezer
CAS Number: 12707-58-3

Order Now

Specification Test Results

Source/Host Isolated from bovine brain.
Purity 98% (TLC)
Solubility Soluble in water (micellar aggregates) or chloroform:methanol (2:1).
Formulation Lyophilized.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR