Myxochelin A ≥99% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0470-C100 AG-CN2-0470-M001
102988-392EA 124.62 USD
102988-392 102988-394
Myxochelin A ≥99% (by NMR)
Myxochelin A

Potent inhibitor of human 5-lipoxygenase (5-LO)


Source/Host: Synthetic. Originally isolated from Pyxidicoccus fallax HKI 727.
Purity: >99% (NMR)
Appearance/Color: Solid.
Solubility: Soluble in DMSO, ethanol, methanol, acetone, ethylacetate or chloroform. Insoluble in water or hexane.


This enzyme catalyzes two initial steps in the conversion of arachidonic acid into leukotrienes, well known mediators of inflammatory and allergic reactions. Iron-chelating compound. Anticancer antibiotic. Shown to inhibit tumor cell invasion in vitro. Antibacterial compound. Antioxidant with free radical scavenging activities.

Storage Temperature: Freezer
CAS Number: 120243-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR