Ledol ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0406-M500 AG-CN2-0406-G001
102988-154EA 55.38 USD
102988-154 102988-156
Ledol ≥95% (by NMR)
Ledol

Toxic sesquiterpenoid. Exhibits expectorant and antitussive effect. Reported to increase blood pressure. Antifungal compound.

Formula: C₁₅H₂₆O
Storage Temperature: Freezer
CAS Number: 577-27-5

Order Now

Specification Test Results

Source/Host Isolated from Ledum palustre.
Purity 95% (NMR)
Appearance/Color White crystalline powder.
Solubility Soluble in DMSO, ethanol or dimethylformamide. Sparingly soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR