Asperphenamate ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Anabellamide
AG-CN2-0171-C250
AG-CN2-0171-M001
102988-142EA
124.62
USD
102988-142
102988-144
Asperphenamate ≥95% (by HPLC)
Asperphenamate
Anticancer compound. Cytotoxic against human breast cancer cells. Induces autophagic cell death in MCF-7 cells. Moderate radical scavenger. Weak acetylcholinesterase (AChE) inhibitor. Shows moderate trypanocidal activity.
Formula:
C₃₂H₃₀N₂O₄ MW: 506.6 g/mol Storage Temperature: Freezer |
CAS Number:
63631-36-7 |
Specification Test Results
Source/Host | Isolated from Aspergillus sp. |
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in ethanol, methanol or DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...