epi-Aszonalenin A ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Epiaszonalenin A

AG-CN2-0163-M001 AG-CN2-0163-M005
102987-430EA 214.62 USD
102987-430 102987-432
epi-Aszonalenin A ≥95% (by HPLC)
epi-Aszonalenin A

Derivative of aszonalenin, a substance P inhibitor.

Formula: C₂₅H₂₅N₃O₃
MW: 415.4 g/mol
Storage Temperature: Freezer
CAS Number: 908853-14-5

Order Now

Specification Test Results

Source/Host Isolated from Neosartorya fischeri.
Purity 95% (HPLC)
Appearance/Color White solid.
Solubility Soluble in DMSO, methanol or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR