NG 012 ≥95% (by TLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Orbuticin derivative, (3R,7S,11R,17R,21R)-7-(Hydroxymethyl)-3,11,17,21-tetramethyl-13,14:23,24-bis[1,3]butadieno-25,27,29,31-tetrahydroxy-2,6,10,16,20-pentaoxacyclotetracosa-13,23-diene-1,5,9,15,19-pentone
AG-CN2-0155-M001
AG-CN2-0155-M005
102988-102EA
138.46
USD
102988-102
102988-104
NG 012 ≥95% (by TLC)
NG 012
Antibiotic
Source/Host: Isolated from Penicillium sp. FKI-5387.
Purity: >95% (TLC)
Appearance/Color: Pale white solid.
Solubility: Soluble in methanol, ethanol, ethyl acetate, chloroform, acetonitrile or acetone. Insoluble in water or hexane.
Antifungal. Nerve growth factor (NGF) potentiator.
Formula:
C₃₂H₃₈O₁₅ MW: 662.7 g/mol Storage Temperature: Freezer |
CAS Number:
141731-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...