Macrosphelide A ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0152-M001 AG-CN2-0152-M005
102988-094EA 193.85 USD
102988-094 102988-096
Macrosphelide A ≥95% (by HPLC)
Macrosphelide A

Antibiotic


Source/Host: Isolated from Paraconiothyrium sporulosum FO-5050.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in methanol, DMSO, chloroform, ethyl acetate or dimethyl ether. Insoluble in water or hexane.


Cell-cell adhesion inhibitor. Antimetastatic compound. Antifungal. Antimicrobial.

Formula: C₁₆H₂₂O₈
Storage Temperature: Freezer
CAS Number: 172923-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR