Harzianopyridone ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
4-Hydroxy-5,6-dimethoxy-3-[(2S,4E)-2-methyl-1-oxo-4-hexen-1-yl]-2(1H)-pyridinone, (-)-Harzianopyridone
Antibiotic
Source/Host: Isolated from Trichoderma harzianum FKI-1509.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO, methanol, ethyl acetate, acetone, acetonitrile or chloroform. Insoluble in water or hexane.
Antifungal and antibacterial compound. Specific mitochondrial complex II (succinate ubiquinone oxidoreductase; succinate dehydrogenase) inhibitor. Herbicidal activity. Anthelmintic compound. Inhibits NADH-fumarate reductase activity of adult Ascaris suum mitochondria.
Formula:
C₁₄H₁₉NO₅ MW: 281.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD23160296 CAS Number: 137813-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...