Aurodox ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
AG-CN2-0133-M001
AG-CN2-0133-MM25
102988-024EA
207.69
USD
102988-024
102988-026
Aurodox ≥95% (by HPLC)
Aurodox
Antibiotic Protein biosynthesis inhibitor by binding to elongation factor Tu (EF-Tu)
Source/Host: Isolated from Streptomyces sp. K06-0806.
Purity: >95% (HPLC)
Appearance/Color: Yellow solid.
Solubility: Soluble in methanol or chloroform. Insoluble in water.
Type 3 secretion system (T3SS) inhibitor. Growth promotor in poultry.
Formula:
C₄₄H₆₂N₂O₁₂ MW: 811.0 g/mol Storage Temperature: Freezer |
CAS Number:
12704-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...