Viridiol ≥95%

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0126-C250 AG-CN2-0126-M001
102987-998EA 83.08 USD
102987-998 102988-000
Viridiol ≥95%
Viridiol

Steroidal antibiotic


Source/Host: Isolated from Trichoderma sp.
Purity: >95% (HPLC)
Appearance/Color: Off-white solid.
Solubility: Soluble in DMSO.


Belongs to the Viridin family, including Wortmannin. Antifungal. Phytotoxin. Shows necrotic activity on plants. Phosphatidylinositol 3-kinase (PI3K) inhibitor.

Storage Temperature: Freezer
CAS Number: 23820-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR