Amidepsine A ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0109-M001 AG-CN2-0109-MM25
102987-378EA 318.46 USD
102987-378 102987-380
Amidepsine A ≥95% (by HPLC)
Amidepsine A

Diacylglycerol acyltransferase (DGAT) inhibitor. Excessive accumulation of triacetylglycerol produced by DGAT may cause fatty liver, obesity and hypertriglyceridemia, which may lead to atherosclerosis, diabetes and metabolic disorders.

Formula: C₂₉H₂₉NO₁₁
Storage Temperature: Freezer
CAS Number: 169181-28-6

Order Now

Specification Test Results

Source/Host Isolated from Humcola sp. FO-2942.
Purity 95% (HPLC)
Appearance/Color Pale white powder.
Solubility Soluble in methanol, benzene or ethyl acetate. Insoluble in water or hexane.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR