Malformin C 95

Supplier: ADIPOGEN CORP MS
Warning

AG-CN2-0107-C250 AG-CN2-0107-M001
102987-370EA 152.31 USD
102987-370 102987-372
Malformin C 95
Malformin C

Peptide antibiotic


Source/Host: Synthetic. Originally isolated from Aspergillus niger FKI-2342.
Purity: >95% (HPLC)
Appearance/Color: White solid.
Solubility: Soluble in DMSO, methanol, ethanol or chloroform. Insoluble in water.


Induces root curvature and malformation in plants. Antibacterial, antimalarial and antitrypanosomal. Disrupts the cell cycle at the G2 checkpoint of cancer cells, leading to sensitization of the cancer cells to anti-cancer reagents.

Formula: C₂₃H₃₉N₅O₅S₂
MW: 529.7 g/mol
Storage Temperature: Freezer
CAS Number: 59926-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR