Atpenin A5 ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Danger
|
Antibiotic
Source/Host: Synthetic. Originally isolated from Penicillium sp. strain FO-125.
Purity: >95% (HPLC)
Appearance/Color: White to off-white solid.
Solubility: Soluble in acetone, acetonitrile, chloroform, ethyl acetate, DMSO, methanol or ethanol. Insoluble in water or hexane.
Antifungal. Potent and specific mitochondrial complex II (succinate-ubiquinone oxidoreductase) inhibitor. Mitochondrial ATP-sensitive potassium (mK(ATP)) channel activator. Cardioprotective. Modulates mitochondrial ROS generation during cardioprotection.
Storage Temperature:
Freezer |
CAS Number:
119509-24-9 UN: 3462 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...