Sauchinone ≥98% (by HPLC)
Supplier: ADIPOGEN CORP MS
Synonyms:
1′-Epimer dihydrocarpanone, (5aR,5a1R,7R,8S,8aR,14aS)-7,8-Dimethyl-5a,5a1,6,7,8,8a-hexahydro-5H-benzo[kl]bis([1,3]dioxolo)[4,5-b:4',5'-g]xanthen-5-one
AG-CN2-0032-M001
AG-CN2-0032-M005
102987-846EA
41.54
USD
102987-846
102987-848
Sauchinone ≥98% (by HPLC)
Sauchinone
Hepatoprotective antioxidant. Anti-inflammatory. Suppresses NF-kB p65 activity. Neuroprotective. Apoptosis inhibitor. Bone resorption/osteoclastogenesis inhibitor. Reduces ROS formation. Antiseptic. AMP-activated kinase (AMPK) activator. Akt phosphorylation inhibitor.
Formula:
C₂₀H₂₀O₆ MW: 356.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD08702698 CAS Number: 177931-17-8 |
Specification Test Results
Source/Host | Isolated from Saururus chinensis. |
Purity |
|
Appearance/Color | White to off-white powder. |
Solubility | Soluble in DMSO or chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...