n-Dodecyl-α-D-maltoside (by HPLC), Ultrapure
Supplier: ADIPOGEN CORP MS
Synonyms:
DDM
AG-CC1-0006-G001
AG-CC1-0006-G005
AG-CC1-0006-G025
102987-832EA
166.15
USD
102987-832
102987-834
102987-836
n-Dodecyl-α-D-maltoside (by HPLC), Ultrapure
n-Dodecyl-alpha-D-maltoside
Non-ionic detergent. For functional solubilization, purification and crystallography of membrane proteins. For the stabilization and activation of enzymes.
Formula:
C₂₄H₄₆O₁₁ MW: 510.6 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00133592 CAS Number: 116183-64-3 |
Specification Test Results
Purity | > 99.5% (HPLC)[beta-Isomer <7% (HPLC)][n-Dodecanol <0.002% (HPLC)] |
Appearance/Color | White powder. |
Solubility | Soluble in water or 100% ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...