Nilotinib

Supplier: ADIPOGEN CORP MS

SYN-1057-M001 SYN-1057-M005 SYN-1057-M010 SYN-1057-M050
102991-692EA 74.77 USD
102991-692 102991-694 102991-696 102991-698
Nilotinib
Nilotinib

Nilotinib is an orally bio-available aminopyrimidine-derivative Bcr-Abl tyrosine kinase inhibitor (IC(50) 30nM) with anti-neoplastic activity.

Formula: C₂₈H₂₂F₃N₇O
MDL Number: MFCD09833716
CAS Number: 641571-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR