N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥95%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Gefitinib
SYN-1042-M001
SYN-1042-M005
SYN-1042-M010
SYN-1042-M050
102991-574EA
59.54
USD
102991-574
102991-576
102991-578
102991-580
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥95%
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine
Gefitinib is a selective, orally available epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor. In NR6wtEGFR and NR6W cells, it inhibits Tyr1173, Tyr992, Tyr1173 and Tyr992 with IC(50) values of 37nM, 37nM, 26nM and 57nM, respectively.
Formula:
C₂₂H₂₄ClFN₄O₃ MW: 446.91 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD04307832 CAS Number: 184475-35-2 |
Specification Test Results
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...