Tropolone tosylate

Supplier: ADIPOGEN CORP MS

CDX-T0085-GG25 CDX-T0085-G025
102991-100EA 220.15 USD
102991-100 102991-102
Tropolone tosylate
Tropolone tosylate

Building block for synthesis.

Formula: C₁₄H₁₂O₄S
MDL Number: MFCD00671574
CAS Number: 38768-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR