2,3,3-Trimethylindolenine

Supplier: ADIPOGEN CORP MS

CDX-T0080-G005 CDX-T0080-G050
102991-086EA 54 USD
102991-086 102991-088
2,3,3-Trimethylindolenine
2,3,3-Trimethylindolenine

Indole derivative used in the preparation of cyanine dye labelleing reagents and other imaging agents.

Formula: C₁₁H₁₃N
MW: 159.23 g/mol
MDL Number: MFCD00005724
CAS Number: 1640-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR