Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Supplier: ADIPOGEN CORP MS
Synonyms:
Tris(3,5-bis(trifluoromethyl)phenyl)phosphine
CDX-T0049-M500
CDX-T0049-GG25
102991-078EA
175.85
USD
102991-078
102991-080
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
Tris[3,5-bis(trifluoromethyl)phenyl]phosphine
A phosphine with enhanced solubility in solvents. Phosphines are used as ligands for many metal complexes.
Formula:
C₂₄H₉F₁₈P MW: 670.28 g/mol |
MDL Number:
MFCD00042428 CAS Number: 175136-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...