JC-1 iodide salt

Supplier: ADIPOGEN CORP MS

Synonyms: 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethylbenzimidazolocarbocyanine iodide, JC-1, 5,5′,6,6′-Tetrachloro-1,1′,3,3′-tetraethyl-imidacarbocyanine iodide, (E)-5,6-Dichloro-2-(3-(5,6-dichloro-1,3-diethyl-1,3-dihydro-2H-benzo[d]imidazol-2-ylidene)prop-1-en-1-yl)-1,3-diethyl-1H-benzo[d]imidazol-3-ium iodide, CBIC2(3), 1,1',3,3'-tetraethyl-5,5',6,6'-tetrachloroimidacarbocyanine iodide

CDX-T0046-M010 CDX-T0046-M100
102991-074EA 404.31 USD
102991-074 102991-076
JC-1 iodide salt
JC-1 iodide salt

A dual-emission potential-sensitive probe that can be used to measure mitochondrial membrane potential. JC-1 is a green-fluorescent monomer at low membrane potential. At higher potentials, JC-1 forms red-fluorescent "J-aggregates," which exhibit broad excitation and very narrow emission spectra. The ratio of red to green fluorescence of JC-1 is dependent only on membrane potential, and not influenced by mitochondrial size, shape, or density.

Formula: C₂₅H₂₇Cl₄IN₄
MW: 652.23 g/mol
MDL Number: MFCD00188475
CAS Number: 47729-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR