2,3,5-Triphenyltetrazolium chloride (TTC) ≥99%

Supplier: ADIPOGEN CORP MS

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

CDX-T0026-G025 CDX-T0026-G100
102991-042EA 138.46 USD
102991-042 102991-044
2,3,5-Triphenyltetrazolium chloride (TTC) ≥99%
2,3,5-Triphenyltetrazolium chloride (TTC)

Triphenyl tetrazolium chloride, TTC, or simply tetrazolium chloride is a redox indicator commonly used in biochemical experiments especially to indicate cellular respiration.


  • Off-white to yellow powder

Formula: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Melting Pt: 235
MDL Number: MFCD00011963
CAS Number: 298-96-4
Merck Index: 13,09815

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR