7-Hydroxy-4-(trifluoromethyl)coumarin

Supplier: ADIPOGEN CORP MS

CDX-T0003-G001 CDX-T0003-GG25 CDX-T0003-G025
102991-022EA 123.23 USD
102991-022 102991-024 102991-026
7-Hydroxy-4-(trifluoromethyl)coumarin
7-Hydroxy-4-(trifluoromethyl)coumarin

This compound is widely used as a substrate for Cytochrome P450. It has also been used as a marker substrate for assessment of Drug-Drug interactions to multiple isoforms of UDP-glucuronosyltransferases. Suitable as pH-indicator. Trifluoromethylumbelliferone is a slightly longer wavelength analog of 4-methylcoumarin (4-MU) with a pKa that more closely matches physiological pH values.

Formula: C₁₀H₅F₃O₃
MDL Number: MFCD00037578
CAS Number: 575-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR