Disodium mesoxalate monohydrate
Supplier: ADIPOGEN CORP MS
CDX-S0028-G010
CDX-S0028-G025
CDX-S0028-G125
102990-976EA
88.62
USD
102990-976
102990-978
102990-980
Disodium mesoxalate monohydrate
Disodium mesoxalate monohydrate
Building block for synthesis.
Formula:
C₃Na₂O₅·1H₂O |
CAS Number:
31635-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...