Sulforhodamine 101
Supplier: ADIPOGEN CORP MS
Synonyms:
Sulforhodamine 101, Texas Red, SR 101, Sulforhodamine 640, 9-(2,4-Disulphophenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium inner salt
Sulforhodamine 101 is a non-fixable red fluorescent dye used as a specific marker for astrocytes and an activity-dependent probe for monitoring regulated exocytosis. In addition to labeling astrocytes, it also labels myelinating oligodendrocytes. Has been reported to affect excitability of neurons and should therefore be used with caution. Key precursor used for preparing rhodamine 101 sulfonyl chloride (also called Texas Red). Spectral data: lambdaex 586 nm| lambdaem 605 nm in H2O and lambdaex 582-588 nm| lambdaem 601-609 nm in pH7 buffer.
Formula:
C₃₁H₃₀N₂O₇S₂ MW: 606.708 g/mol |
MDL Number:
MFCD00012407 CAS Number: 60311-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...