Sulforhodamine 101 (free acid)

Supplier: ADIPOGEN CORP MS

CDX-S0008-M005 CDX-S0008-M100
102990-968EA 148.15 USD
102990-968 102990-970
Sulforhodamine 101 (free acid)
Sulforhodamine 101 acid chloride

Sulforhodamine 101 sulfonyl chloride (also called Texas Red sulfonyl chloride) is a popular long wavelength amine-reactive rhodamine dye. Because the dye is quite unstable in water, reactions with amines in an aqueous solution must be carried out at low temperature (over ice or at 4°C) and usually at pH 8.5.

Formula: C₃₁H₂₉ClN₂O₆S₂
MDL Number: MFCD00012406
CAS Number: 82354-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR