Resveratrol
Supplier: ADIPOGEN CORP MS
Synonyms:
trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
Selective inhibitor of COX−1. Resveratrol is a phenolic phytoalexin found in grape skin and other plants. It has intracellular antioxidant activity and activates SIRT1, a NAD+-dependent histone deacetylase involved in mitochondrial biogenesis and the enhancement of peroxisome proliferator-gamma-activated receptor coactivator-1alpha (PGC-1alpha) and FOXO activity. The anti-diabetic, neuroprotective and anti-adipogenic actions of resveratrol may be mediated via SIRT1 activation.
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol |
MDL Number:
MFCD00133799 CAS Number: 501-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...