Methyl 2-(chlorocarbonyl)benzoate ≥96% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: Methyl phthaloyl chloride

CDX-P0105-G001 CDX-P0105-GG25 CDX-P0105-G010
102990-828EA 131.54 USD
102990-828 102990-830 102990-832
Methyl 2-(chlorocarbonyl)benzoate ≥96% (by HPLC)
Methyl-2-(chlorocarbonyl)benzoate

Building block for synthesis.

Formula: C₉H₇ClO₃
MW: 198.61 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11656039
CAS Number: 4397-55-1
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 96% (HPLC)
Appearance/Color Solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR