1-Pyrenecarboxaldehyde

Supplier: ADIPOGEN CORP MS

CDX-P0093-G020 CDX-P0093-G100
102990-818EA 225.69 USD
102990-818 102990-820
1-Pyrenecarboxaldehyde
1-Pyrenecarboxaldehyde

Building block for synthesis.

Formula: C₁₇H₁₀O
MDL Number: MFCD00004139
CAS Number: 3029-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR