Phenol red sodium salt

Supplier: ADIPOGEN CORP MS

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

CDX-P0026-G001 CDX-P0026-G010 CDX-P0026-G050
102990-802EA 34.62 USD
102990-802 102990-804 102990-806
Phenol red sodium salt
Phenol red sodium salt

Water soluble pH indicator used in the 6.8 (yellow) to 8.2 (red) range and frequently used in cell biology laboratories. Widely used as cell tissue culture pH marker. A small amount of phenol red added to the growth medium will have a pink-red color under normal conditions. Typically, 15 mg/l are used for cell culture. In the event of problems, waste products produced by dying cells or overgrowth of contaminants will cause a change in pH, leading to a change in indicator color. For example, a culture of relatively slowly dividing mammalian cells can be quickly overgrown by bacterial contamination. This usually results in an acidification of the medium, turning it yellow. The waste products produced by the mammalian cells themselves will slowly decrease the pH, gradually turning the solution orange and then yellow. This color change is an indication that even in the absence of contamination, the medium needs to be replaced. Shown to be a weak estrogen mimic (possible through inpurities), which can enhance the growth of cells that express the estrogen receptor in cell cultures. Might be useful as a differentiation factor.

Formula: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
MDL Number: MFCD00066901
CAS Number: 34487-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR