1,4-Phenylene diisothiocyanate

Supplier: ADIPOGEN CORP MS

CDX-P0022-G005 CDX-P0022-G020 CDX-P0022-G100
102990-790EA 113.54 USD
102990-790 102990-792 102990-794
1,4-Phenylene diisothiocyanate
1,4-Phenylene diisothiocyanate

Reagent for solid-phase sequencing.

Formula: C₈H₄N₂S₂
MDL Number: MFCD00004811
CAS Number: 4044-65-9
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR