1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 1,3,6,8-Pyrenetetrasulphonic acid tetrasodium salt

CDX-P0021-GG25 CDX-P0021-G010 CDX-P0021-G100
102990-784EA 157.85 USD
102990-784 102990-786 102990-788
1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt ≥98% (by HPLC)
1,3,6,8-Pyrenetetrasulfonic acidtetrasodium salt

Fluorescent probe and pH indicator. It fluoresces blue in acidic solutions and in acidic organelles but fluoresces green in more basic organelles. This compound is potentially useful as a coloring agent, biological stain or optical detecting reagent. It is used in water-in-oil-in-water emulsions and as a substrate for lipase activity. Spectral data: lambdaex 375nm, lambdaem 404nm in water.

Formula: C₁₆H₆Na₄O₁₂S₄
MW: 610.411 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09263184
CAS Number: 59572-10-0

Order Now

Specification Test Results

Source/Host Synthetic
Purity >98% (HPLC)
Appearance/Color White, yellow crystalline.
Solubility Soluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR