Oxyresveratrol

Supplier: ADIPOGEN CORP MS

Synonyms: 2,3',4,5'-Tetrahydroxy-trans-stilbene, (E)-4-(3,5-Dihydroxystyryl)benzene-1,3-diol

CDX-O0035-M100 CDX-O0035-G001
102990-690EA 210.46 USD
102990-690 102990-692
Oxyresveratrol
Oxyresveratrol

Naturallly occuring analog of resveratrol. Potent antioxidant and free-radical scavenger. Shown to have neuroprotective activity against cerebral ischemia and against traumatic injury. Apoptosis inhibitor in transient cerebral ischemia. Inhibits viral DNA replication and late viral protein synthesis of African Swine fever virus. Has depigmenting effects by effectively inhibiting tyrosinase activity, which catalyzes the rate-limiting step in synthesizing melanin pigments. Has anti-inflammatory properties based on inhibition of iNOS expression through down-regulation of NF-kB binding activity and significant inhibition of COX-2 activity. Antihyperlipidemic agent.

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
MDL Number: MFCD11974969
CAS Number: 29700-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR