Rhodamine B octadecyl ester perchlorate

Supplier: ADIPOGEN CORP MS

CDX-O0022-M020 CDX-O0022-M100
102990-672EA 193.85 USD
102990-672 102990-674
Rhodamine B octadecyl ester perchlorate
Rhodamine B octadecyl ester perchlorate

Sensitive membrane dye used for potassium (fiber-optic sensors) and nitrate sensing and other investigations of membranes. Lipophilic energy transfer acceptor from lipophilic fluoresceins in fluorescence energy transfer (FRET) assays in cell fusion experiments (Ex/Em: 554/575nm in methanol).

Formula: C₄₆H₆₇ClN₂O₇
CAS Number: 142179-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR