Phthalaldehyde ≥99% (by HPLC)

Supplier: ADIPOGEN CORP MS
Danger

CDX-O0008-G001 CDX-O0008-G005 CDX-O0008-G025
102990-662EA 54 USD
102990-662 102990-664 102990-666
Phthalaldehyde ≥99% (by HPLC)
Phthalaldehyde

Flow cytometry measurement tool. Fluorescence probe for glutathione and glutathione disulfide levels, primary amines, peptides and biological thiols. Reacts with primary amines to produce a product that emits a highly fluorescent blue colour. Spectral data: lambdaex 334 nm; lambdaem 455 nm (Thiol Adduct). Used for the quick visualization of histamine, characterized by the appearance of a yellow stain. Used for precolumn derivatization of amino acids for HPLC separation. High-level disinfectant/sterilant for endoscopes, thermometers, rubber and plastic equipment, and medical/surgical instruments that cannot be sterilized by heat, based on its antimicrobial efficacy and low inhalation toxicity.

Formula: C₈H₆O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00003335
CAS Number: 643-79-8
UN: 2923
ADR: 8 (6.1),II

Order Now

Specification Test Results

Source/Host Synthetic
Purity >99% (HPLC)
Appearance/Color Light yellow powder.
Solubility Soluble in water (53 mg/ml), acetic acid (40 mg/ml) or methanol (21 mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR