Nile Red

Supplier: ADIPOGEN CORP MS

Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

CDX-N0107-G001 CDX-N0107-G005
102990-646EA 263.08 USD
102990-646 102990-648
Nile Red
Nile red

Uncharged, hydrophobic, photostable fluorescent probe that strongly fluoresces bright red in hydrophobic (lipid-rich) environments, but is almost nonfluorescent in water. This lipophilic stain is commonly used for the detection of intracellular lipid droplets in cells such as adipocytes by fluorescence microscopy and flow cytometry. It has been shown to detect the exposure or formation of new hydrophobic surfaces induced by ligand binding to calmodulin, oligomerization of melittin, or unfolding of ovalbumin during early thermal denaturation. This product has been employed to stain cell structures, as well as to visualize and localize colloidal drug carriers. Has been used to stain lipophilic proteins that have been separated by SDS-PAGE electrophoresis or to detect sphingolipids on thin-layer chromatograms. The fluorescence is unaffected between pH 4.5 and 8.5 and may be used in dilute concentrations to investigate hydrophobic protein sites. Spectral properties: Intracellular fat vacuoles, filled with neutral lipids such as cholesterol, lipoproteins and triglycerides will fluoresce green (Abs/Em =485/525) while polar lipids, such as phospholipids, will fluoresce red (Abs/Em = 552/636 nm).

Formula: C₂₀H₁₈N₂O₂
MW: 318.38 g/mol
MDL Number: MFCD00011639
CAS Number: 7385-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR