5-Nonanol

Supplier: ADIPOGEN CORP MS

CDX-N0066-L050 CDX-N0066-L250
102990-634EA 70.62 USD
102990-634 102990-636
5-Nonanol
5-Nonanol

Bulding block for synthesis.

Formula: C₉H₂₀O
MW: 144.26 g/mol
MDL Number: MFCD00021944
CAS Number: 623-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR