NBD-PZ (=4-Nitro-7-piperazino-2,1,3-benzoxadiazole)

Supplier: ADIPOGEN CORP MS

Synonyms: 4-Nitro-7-piperazinobenzofurazan, 4-Nitro-7-piperazino-2,1,3-benzoxadiazole

CDX-N0014-M200 CDX-N0014-G001
102990-608EA 225.69 USD
102990-608 102990-610
NBD-PZ (=4-Nitro-7-piperazino-2,1,3-benzoxadiazole)
NBD-PZ (=4-nitro-7-piperazino-2,1,3-benzoxadiazole)

This common fluorescent reagent often used in carbohydrate analysis, is also useful in fluorescence visualization of lysosomes in live cell lines in culture. This fluorophore that has the unusual environmentally sensitive property of increasing fluorescence in low pH environments. Comparison of staining patterns with the common LysoTracker™ or LysoSensor™ dyes indicated comparable staining, albeit using a higher concentration of NBD-piperazine (1μM versus 100nM for the Lyso-Dyes) but at a lower cost per assay. Also used for the determination of mono- and diisocyantes in air samples, and determination of lactate, hydroxybutyrate and other intermediates in body fluids. The piperazino moiety reacts readily with isocyantes to form the corresponding urea-derivatives. The derivatives can be detected by LC with UV or fluorescent-detection. Spectral data: lambdaex 470nm, lambdaem 540nm.

Formula: C₁₀H₁₁N₅O₃
MW: 249.23 g/mol
MDL Number: MFCD00191507
CAS Number: 139332-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR