C12-NBD, free acid ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS

Synonyms: 12-(7-Nitrobenzofurazan-4-ylamino)dodecanoic acid, NBD-dodecanoic acid

CDX-N0013-M100 CDX-N0013-M500
102990-604EA 234 USD
102990-604 102990-606
C12-NBD, free acid ≥98% (by TLC)
C12-NBD, free acid

Suitable for probing the ligand binding sites of fatty acid and sterol carrier proteins as well as the investigation of lipolysis processes.

Formula: C₁₈H₂₆N₄O₅
MW: 378.429 g/mol
Storage Temperature: Freezer
CAS Number: 96801-39-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (TLC)
Appearance/Color Solid.
Solubility Soluble in methanol or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR