3-Methyladenine

Supplier: ADIPOGEN CORP MS

Synonyms: 3-Methyl-3H-purin-6-amine, 3-Methyl-3H-adenine

CDX-M0203-M025 CDX-M0203-M100 CDX-M0203-M250
102990-568EA 62.31 USD
102990-568 102990-570 102990-572
3-Methyladenine
3-Methyladenine

Used to inhibit and study the mechanism of autophagy (lysosomal self-degradation) and apoptosis under various conditions. Inhibits autophagy by blocking autophagosome formation via the inhibition of type III Phosphatidylinositol 3-kinases (PI-3K).

Formula: C₆H₇N₅
MW: 149.16 g/mol
MDL Number: MFCD00010531
CAS Number: 5142-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR