1-Butyn-3-yl p-Toluenesulfonate

Supplier: ADIPOGEN CORP MS

Synonyms: p-Toluenesulphonic acid 1-Butyn-3-yl Ester, (RS)-1-Methyl-2-propynyl-p-toluenesulphonate, But-3-yn-2-yl 4-methylbenzenesulphonate

CDX-M0137-G001 CDX-M0137-G005
102990-546EA 88.62 USD
102990-546 102990-548
1-Butyn-3-yl p-Toluenesulfonate
1-Butyn-3-yl-p-toluenesulfonate

Alkynes building blocks for cross-coupling reactions.

Formula: C₁₁H₁₂O₃S
MW: 224.27 g/mol
MDL Number: MFCD00060095
CAS Number: 53487-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR