Methyl 5-aminonicotinate ≥96% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
Methyl 5-aminonicotinate, Methyl 5-aminopyridine-3-carboxylate
CDX-M0129-G001
CDX-M0129-GG25
CDX-M0129-G010
102990-518EA
127.38
USD
102990-518
102990-520
102990-522
Methyl 5-aminonicotinate ≥96% (by HPLC)
Methyl-5-aminonicotinate
Heterocyclic building block.
Formula:
C₇H₈N₂O₂ MW: 152.1500 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04038688 CAS Number: 36052-25-2 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...