Methyl-6-hydroxy-5-nitronicotinate ≥96%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
Methyl 6-hydroxy-5-nitronicotinate, Methyl 6-hydroxy-5-nitropyridine-3-carboxylate
CDX-M0128-G001
CDX-M0128-G010
102990-514EA
156.46
USD
102990-514
102990-516
Methyl-6-hydroxy-5-nitronicotinate ≥96%
Methyl-6-hydroxy-5-nitronicotinate
Heterocyclic building block.
Formula:
C₇H₆N₂O₅ Storage Temperature: Refrigerator |
MDL Number:
MFCD10699641 CAS Number: 222970-61-8 UN: 3261 ADR: 8,III |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...