cis-11-Methyl-2-dodecenoic acid

Supplier: ADIPOGEN CORP MS

Synonyms: (Z)-11-Methyldodec-2-enoic acid, DSF, Diffusible signal factor

CDX-M0075-M010 CDX-M0075-M100
102990-434EA 195.23 USD
102990-434 102990-436
cis-11-Methyl-2-dodecenoic acid
cis-11-Methyl-2-dodecenoic acid

A diffusible signal factor (DSF) in extracellular microbial and fungal communication systems. In a DSF bioassay, the minimum concentration of cis-Delta2-11-methyl-dodecenoic acid required for induction of a DSF biosensor was about 0.5 µM, which is 200-fold less than that of the conformational isomer (trans-Delta2-11-methyl-dodecenoic acid) and 20,000-fold lower than that of the corresponding saturated fatty acid (11-methyl-dodecanoic acid).

Formula: C₁₃H₂₄O₂
MW: 212.33 g/mol
CAS Number: 677354-23-3
UN: 1231
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR