N-Succinimidyl 7-Methoxycoumarin-3-carboxylate

Supplier: ADIPOGEN CORP MS

Synonyms: 7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 7-methoxy-2-oxo-2H-chromene-3-carboxylate

CDX-M0017-M025 CDX-M0017-M125
102990-376EA 181.38 USD
102990-376 102990-378
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate

Excellent amino-reactive tag that has strong blue fluorescence. Can be used to create blue-fluorescent bioconjugates with an excitation/emission spectra of ~358/410 nm. Used to label neuropeptides, peptides, nucleotides and cells.

Formula: C₁₅H₁₁NO₇
MDL Number: MFCD00467593
CAS Number: 150321-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR