7-Methoxycoumarin-3-carboxylic Acid

Supplier: ADIPOGEN CORP MS

CDX-M0016-M500 CDX-M0016-G005
102990-372EA 202.15 USD
102990-372 102990-374
7-Methoxycoumarin-3-carboxylic Acid
7-Methoxycoumarin-3-carboxylic acid

An amine-reactive blue fluorescent probe that provides a distinguishable contrast when used with longer-wavelength probes in certain multicolor fluorescence applications such as nucleic acid and protein microarrays, in situ hybridization and immunofluorescence. This carboxylic acid derivative of 7-methoxycoumarin can be used for coupling to amines, hydrazines or hydroxylamines on biomolecules of interest. Spectral data: Ex=336nm, Em=402nm (Buffer, pH9).

Formula: C₁₁H₈O₅
MDL Number: MFCD00452770
CAS Number: 20300-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR